Assay Details
Gene Symbol: 4930555O08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016136.1 | - | 1 - 2 | 254 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | F-box and WD-40 domain protein 11 | Fbxw11 |
2310065A07Rik, AA536858, BTRC2, BTRCP2, Fbxw1b, HOS |
-42.9kb | Anti-sense | Chr. 11 - 32642003 - 32746816 on NCBI Build GRCm38 |
Downstream | small integral membrane protein 23 | Smim23 | 1700008A04Rik | 29.7kb | Sense | Chr. 11 - 32820376 - 32824594 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 222834 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691015 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09622163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09242429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09703582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09242430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09703577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08991770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09703580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09625349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690950 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690988 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09195632 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09376089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690958 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09060603 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690941 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690956 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09239479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691030 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09219755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09043113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09043107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09622168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09043111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09242427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690913 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09622165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09566008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690984 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27690915 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27691020 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 660862 | - | - |
