Assay Details
| Gene Symbol: C330013F16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045456.1 | 1 - 2 | 132 | 79 | ||
| GenBank mRNA | AK021198.1 | - | 1 - 2 | 132 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

C330013F16Rik
Genomic Map