Assay Details
| Gene Symbol: Gm27162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039836.1 | - | 1 - 2 | 81 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 3331 | Gm3331 | -78.3kb | Anti-sense | Chr. 9 - 32783146 - 32833574 on NCBI Build GRCm38 | |
| Downstream | predicted gene 27166 | Gm27166 | 102.8kb | Anti-sense | Chr. 9 - 33031791 - 33090799 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm27162
Genomic Map