Assay Details
| Gene Symbol: Gm27162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039836.1 | - | 3 - 4 | 376 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 3331 | Gm3331 | -73.2kb | Anti-sense | Chr. 9 - 32783146 - 32833574 on NCBI Build GRCm38 | |
| Downstream | predicted gene 27166 | Gm27166 | 120.7kb | Anti-sense | Chr. 9 - 33031791 - 33090799 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787573 | AK012733.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787573 | AK147090.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787573 | BC022595.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787573 | BC052439.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787573 | NM_029581.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419555 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34723 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06744343 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06744311 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01900829 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07463432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07547945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07463413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07702757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07670625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08029191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07463425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07670605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07463421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07918029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07670608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07670615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07702759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07463443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07718030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07760318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07670618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07670624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08029189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07922480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07754933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07704261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08081441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07942999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07670607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07991832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07704263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08141551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07670627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07918024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26408581 | - | - |

Gm27162
Genomic Map