Assay Details
| Gene Symbol: A330043C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039441.1 | - | 1 - 2 | 144 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13652 | Gm13652 | OTTMUSG00000013241 | -8.8kb | Sense | Chr. 2 - 75126034 - 75138356 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9430019J16 gene | 9430019J16Rik | 258.4kb | Sense | Chr. 2 - 75436057 - 75444179 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121924 | AK039441.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139198 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788567 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | AK004708.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | AK010448.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | AK034740.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | AK049132.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | AK169377.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | AK169527.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | AK191314.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | AK220207.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | BC028870.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | BC068122.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | BC087940.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | XM_001479473.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738423 | XM_001479496.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 2840 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121924 | AK039441.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00406887 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00406836 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04569900 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00406887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01609955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01801475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01583249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341998 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02116567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01801454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01253912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01309751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01539631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01253905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02270292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02270269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01424036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01309755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01609948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01253906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01253900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01253898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01270823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01398310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21341997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21342161 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 396680 | - | - |

A330043C09Rik
Genomic Map