Assay Details
| Gene Symbol: Gm12829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050995.1 | - | 109 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TraB domain containing 2B | Trabd2b |
Gm12824, Hkat, OTTMUSG00000008561 |
-65.7kb | Sense | Chr. 4 - 114406194 - 114615098 on NCBI Build GRCm38 |
| Downstream | predicted gene 12830 | Gm12830 | OTTMUSG00000008584 | 135.9kb | Sense | Chr. 4 - 114821720 - 114856166 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12829
Genomic Map