Assay Details
| Gene Symbol: LOC101056093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052180.1 | - | 1 - 2 | 1013 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1110002J07 gene | 1110002J07Rik | -286.0 | Anti-sense | Chr. 10 - 66912191 - 66920616 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31763 |
Gm31763 | 81.5kb | Sense | Chr. 10 - 67005075 - 67041326 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

LOC101056093
Genomic Map