Assay Details
| Gene Symbol: Gm11335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006302.1 | - | 1 - 2 | 269 | 151 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
butyrophilin, subfamily 2, member A2 |
Btn2a2 |
Btn2, D030063K05 |
-38.2kb | Sense | Chr. 13 - 23477049 - 23488858 on NCBI Build GRCm38 |
| Downstream |
histone cluster 1, H4h |
Hist1h4h | 1700024H08Rik | 101.0 | Anti-sense | Chr. 13 - 23531044 - 23531478 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm11335
Genomic Map