Assay Details
| Gene Symbol: Gm15346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC027664.1 | - | 1 - 2 | 91 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
polymerase (DNA directed), beta |
Polb | A430088C08Rik | -32.8kb | Anti-sense | Chr. 8 - 22628119 - 22653468 on NCBI Build GRCm38 |
| Downstream |
plasminogen activator, tissue |
Plat |
AU020998, AW212668, D8Ertd2e, tPA |
62.1kb | Sense | Chr. 8 - 22757744 - 22782848 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15346
Genomic Map