Assay Details
| Gene Symbol: 4930597A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016402.1 | - | 1 - 2 | 33 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 145 | Rnf145 |
3732413I11Rik, TMRF1 |
-20.8kb | Sense | Chr. 11 - 44518680 - 44565520 on NCBI Build GRCm38 |
| Downstream | early B cell factor 1 | Ebf1 |
Ebf, O/E-1, OE-1, Olf-1, Olf1 |
30.6kb | Sense | Chr. 11 - 44618100 - 45008096 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930597A21Rik
Genomic Map