Assay Details
| Gene Symbol: 2900052L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013668.1 | - | 1 - 2 | 97 | 109 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY712645.1 | - | 96 | Chr. - - [-] on Build | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 11770 | Gm11770 | OTTMUSG00000004111 | -7.7kb | Sense | Chr. 11 - 120213034 - 120222100 on NCBI Build GRCm38 |
| Downstream | BAH domain and coiled-coil containing 1 | Bahcc1 |
B930044J06, BC060615 |
1.3kb | Anti-sense | Chr. 11 - 120232904 - 120292297 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013668.1 transcripts.
Set Membership: |
|

2900052L18Rik
Genomic Map