Assay Details
| Gene Symbol: Gm11762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045099.1 | 1 - 2 | 1136 | 91 | ||
| GenBank mRNA | AK053782.1 | - | 1 - 2 | 1139 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 287377 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765366 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112565 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02444744 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02444755 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02444733 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104380 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02444755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09518409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09465255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09660733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09454489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09196612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09558311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09454487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09196617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09186966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09186948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09186949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09401258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09255608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28178728 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1100019242.mm.1 | NR_045099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019242.mm.1 | NR_045099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019242.mm.1 | TC1100002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019240.mm.1 | NR_045099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019240.mm.1 | NR_045099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019240.mm.1 | TC1100002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019241.mm.1 | NR_045099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019241.mm.1 | NR_045099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019241.mm.1 | TC1100002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019243.mm.1 | NR_045099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019243.mm.1 | NR_045099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019243.mm.1 | TC1100002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019244.mm.1 | NR_045099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019244.mm.1 | NR_045099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019244.mm.1 | TC1100002006.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 675013 | - | - |

Gm11762
Genomic Map