Assay Details
| Gene Symbol: Gm20750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040555.1 | 4 - 5 | 393 | 79 | ||
| GenBank mRNA | AK034478.1 | - | 4 - 5 | 393 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30292 |
Gm30292 | -108.9kb | Sense | Chr. 3 - 52789004 - 52797807 on NCBI Build GRCm38 | |
| Downstream | component of oligomeric golgi complex 6 | Cog6 |
4933405E16Rik, AU018618, mKIAA1134 |
71.6kb | Sense | Chr. 3 - 52982123 - 53017223 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm20750
Genomic Map