Assay Details
| Gene Symbol: Gm38509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110488.1 | 1 - 2 | 216 | 83 | ||
| GenBank mRNA | AW988896.1 | - | 1 - 2 | 216 | 83 | |
| BC051220.1 | - | 1 - 2 | 225 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 3143 | Gm3143 | -55.4kb | Anti-sense | Chr. 3 - 34699640 - 34716662 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 21388 |
Gm21388 | 44.2kb | Sense | Chr. 3 - 34826557 - 34874076 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm38509
Genomic Map