Assay Details
Gene Symbol: Gm33602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK078946.1 | - | 2 - 3 | 1186 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930402K13 gene | 4930402K13Rik | -73.1kb | Anti-sense | Chr. X - 9104562 - 9106342 on NCBI Build GRCm38 | |
Downstream | LanC lantibiotic synthetase component C-like 3 (bacterial) | Lancl3 | 6030463G20Rik | 12.6kb | Anti-sense | Chr. X - 9199973 - 9268085 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
