Assay Details
| Gene Symbol: Gm32556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040580.1 | - | 1 - 2 | 111 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ras homolog gene family, member H |
Rhoh |
5830400A04Rik, AU019774, Arhh |
-17.2kb | Anti-sense | Chr. 5 - 65861399 - 65896700 on NCBI Build GRCm38 |
| Downstream |
cholinergic receptor, nicotinic, alpha polypeptide 9 |
Chrna9 |
2410015I05Rik, Acra9, EG666827, Gm8311 |
15.1kb | Anti-sense | Chr. 5 - 65934764 - 65977560 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm32556
Genomic Map