Assay Details
| Gene Symbol: Gm40289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030193.1 | - | 1 - 2 | 306 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 34055 |
Gm34055 | -85.8kb | Sense | Chr. 5 - 37863146 - 37897934 on NCBI Build GRCm38 | |
| Downstream | syntaxin 18 | Stx18 |
1810035L21Rik, 4933425D03Rik, AU041562 |
52.6kb | Sense | Chr. 5 - 38038622 - 38137770 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm40289
Genomic Map