Assay Details
| Gene Symbol: 1110058D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004294.1 | - | 1 - 2 | 107 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | F-box and leucine-rich repeat protein 17 | Fbxl17 |
6330576B01Rik, AI452053, BB073797, C130023C01Rik, Fbx13, Fbxo13 |
-545.8kb | Sense | Chr. 17 - 63045949 - 63504042 on NCBI Build GRCm38 |
| Downstream |
praja 2, RING-H2 motif containing |
Pja2 |
AI447901, AL022700, Neurodap1, mKIAA0438 |
230.3kb | Sense | Chr. 17 - 64281006 - 64331883 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

1110058D11Rik
Genomic Map