Assay Details
Gene Symbol: 2310015A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK009349.1 | - | 1 - 2 | 237 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | salt inducible kinase 1 | Sik1 |
Msk, Sik, Snf1lk |
-52.1kb | Sense | Chr. 17 - 31844250 - 31855792 on NCBI Build GRCm38 |
Downstream |
predicted gene, 30571 |
Gm30571 | 3.5kb | Anti-sense | Chr. 17 - 31912567 - 31913803 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
