Assay Details
| Gene Symbol: Pir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027153.3 | NP_081429.1 | 2 - 3 | 375 | 85 | |
| GenBank mRNA | AK009757.1 | - | 2 - 3 | 318 | 85 | |
| AK082922.1 | - | 2 - 3 | 260 | 85 | ||
| AK090023.1 | - | 2 - 3 | 232 | 85 | ||
| AK134541.1 | - | 2 - 3 | 376 | 85 | ||
| AK138314.1 | - | 2 - 3 | 371 | 85 | ||
| AK151301.1 | - | 2 - 3 | 278 | 85 | ||
| BY718294.1 | - | 2 - 3 | 376 | 85 | ||
| BY720928.1 | - | 2 - 3 | 376 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pir
Genomic Map