Assay Details
| Gene Symbol: Pir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301402.1 | NP_001288331.1 | 5 - 6 | 718 | 116 | |
| NM_027153.3 | NP_081429.1 | 6 - 7 | 847 | 116 | ||
| GenBank mRNA | AK009757.1 | - | 6 - 7 | 790 | 116 | |
| AK134541.1 | - | 6 - 7 | 846 | 116 | ||
| AK138314.1 | - | 6 - 7 | 843 | 116 | ||
| AK151301.1 | - | 6 - 7 | 750 | 116 | ||
| BC024062.1 | - | 6 - 7 | 753 | 5067536 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pir
Genomic Map