Assay Details
| Gene Symbol: 1700061E18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006851.1 | - | 1 - 2 | 256 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | androgen dependent TFPI regulating protein | Adtrp | 9530008L14Rik | Anti-sense | Chr. 13 - 41763148 - 41847616 on NCBI Build GRCm38 | |
| Downstream | predicted gene 10790 | Gm10790 | ENSMUSG00000074927 | 64.1kb | Sense | Chr. 13 - 41865915 - 41913092 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700061E18Rik
Genomic Map