Assay Details
| Gene Symbol: Ppp1r3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029628.1 | NP_083904.1 | 1 - 2 | 74 | 62 | |
| GenBank mRNA | AK079637.1 | - | 1 - 2 | 45 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_029628.1 transcripts.
Set Membership: |
|

Ppp1r3g
Genomic Map