Assay Details
| Gene Symbol: Gm6277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045421.1 | 3 - 4 | 404 | 85 | ||
| GenBank mRNA | AK140316.1 | - | 3 - 4 | 405 | 85 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058448 | AK140316.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058448 | BC030050.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68780 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 792905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64395 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103278 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058448 | AK140316.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058448 | BC030050.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P811150 | BI079333.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13899205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13989834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14056598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13956243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13785523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14056602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13785527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13691559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31497120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14077308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13956249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13899203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Mouse Genome 430 2.0 Array | 1455844_at | - | - |
| Mouse Genome 430 2.0 Array | 1455844_PM_at | - | - |
| Mouse Expression Set 430 | 1455844_at | - | - |
| Mouse Expression Set 430 | 1455844_PM_at | - | - |
| Murine Genome U74v2 Set | 134789_PM_at | - | - |
| Murine Genome U74v2 Set | 134789_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455844_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455844_PM_at | - | - |

Gm6277
Genomic Map