Assay Details
| Gene Symbol: 4933434E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287086.1 | NP_001274015.1 | 2 - 3 | 420 | 58 | |
| GenBank mRNA | BC018438.1 | - | 2 - 3 | 350 | 2609299 | 58 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC018438.1,NM_001287086.1 transcripts.
This assay is in the 5'UTR region of the BC018438.1,NM_001287086.1 transcripts.
Set Membership: |
|

4933434E20Rik
Genomic Map