Assay Details
| Gene Symbol: Rd3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080710.1 | - | 1 - 2 | 71 | ||
| BC048943.1 | - | 2 - 3 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Rd3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080710.1 | - | 1 - 2 | 71 | ||
| BC048943.1 | - | 2 - 3 | 71 | |||
Set Membership: |
|