Assay Details
| Gene Symbol: Gja3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016975.3 | NP_058671.2 | 1 - 2 | 63 | ||
| GenBank mRNA | AY390398.1 | - | 1 - 2 | 63 | ||
| BC138492.1 | - | 1 - 2 | 8861512 | 63 | ||
| BC138493.1 | - | 1 - 2 | 8861513 | 63 | ||
| BY127839.1 | - | 1 - 2 | 63 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY390398.1, BC138492.1, BC138493.1, NM_016975.3 transcripts.
Set Membership: |
|

Gja3
Genomic Map