Assay Details
| Gene Symbol: Med29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026042.3 | NP_080318.2 | 1 - 2 | 237 | 58 | |
| GenBank mRNA | AK005112.1 | - | 1 - 2 | 239 | 58 | |
| AK013006.1 | - | 1 - 2 | 237 | 58 | ||
| BC023192.1 | - | 1 - 2 | 220 | 4947722 | 58 | |
| BC083314.1 | - | 1 - 2 | 219 | 6823222 | 58 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Med29
Genomic Map