Assay Details
Gene Symbol: Zfp942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199048.1 | NP_001185977.1 | 4 - 5 | 616 | 83 | |
GenBank mRNA | BC096765.1 | - | 4 - 5 | 602 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001199048.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33083 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527077 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551999 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935489 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374849 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856885 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119054 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111262 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159930 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45165 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470744 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952004 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497167 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276438 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74775 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15963 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61099 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10448198 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448199 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448197 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448201 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448200 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432887_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454402_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455433_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443456_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10448195 | - | - |
Mouse Genome 430 2.0 Array | 1432887_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1454402_at | - | - |
Mouse Genome 430 2.0 Array | 1443456_at | - | - |
Mouse Genome 430 2.0 Array | 1454402_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1443456_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455433_at | - | - |
Mouse Genome 430 2.0 Array | 1432887_at | - | - |
Mouse Genome 430 2.0 Array | 1455433_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73233_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73233_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700013690.mm.1 | NM_001199048.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013690.mm.1 | TC1700001563.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013691.mm.1 | NM_001199048.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013691.mm.1 | TC1700001563.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013695.mm.1 | NM_001199048.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013695.mm.1 | TC1700001563.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013689.mm.1 | NM_001199048.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013689.mm.1 | TC1700001563.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013693.mm.1 | NM_001199048.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013693.mm.1 | TC1700001563.mm.1 | MTA TC |
Mouse Expression Set 430 | 1455433_at | - | - |
Mouse Expression Set 430 | 1437882_PM_s_at | - | - |
Mouse Expression Set 430 | 1455433_PM_at | - | - |
Mouse Expression Set 430 | 1437882_s_at | - | - |
Mouse Expression Set 430 | 1454402_PM_at | - | - |
Mouse Expression Set 430 | 1443456_PM_at | - | - |
Mouse Expression Set 430 | 1454402_at | - | - |
Mouse Expression Set 430 | 1443456_at | - | - |
Mouse Expression Set 430 | 1432887_PM_at | - | - |
Mouse Expression Set 430 | 1432887_at | - | - |
Murine Genome U74v2 Set | 103710_PM_at | - | - |
Murine Genome U74v2 Set | 103710_at | - | - |
Mouse Genome 430A 2.0 Array | 1437882_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1437882_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1455433_at | - | - |
Mouse Genome 430A 2.0 Array | 1455433_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1455433_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1455433_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454402_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443456_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454402_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432887_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432887_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443456_PM_at | - | - |
