Assay Details
| Gene Symbol: Olfr688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011533.2 | NP_001011533.2 | 1 | 143 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr688| Gene Symbol: Olfr688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011533.2 | NP_001011533.2 | 1 | 143 | 130 | |
Set Membership: |
|