Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600003298.mm.1 |
NM_001123037.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003298.mm.1 |
NM_001123038.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003298.mm.1 |
NM_013506.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003298.mm.1 |
NR_110335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003298.mm.1 |
TC1600000329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003302.mm.1 |
NM_001123037.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003302.mm.1 |
NM_001123038.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003302.mm.1 |
NM_013506.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003302.mm.1 |
NR_110335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003302.mm.1 |
TC1600000329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003297.mm.1 |
NM_001123037.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003297.mm.1 |
NM_001123038.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003297.mm.1 |
NM_013506.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003297.mm.1 |
NR_110335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003297.mm.1 |
TC1600000329.mm.1 |
MTA TC |