Assay Details
| Gene Symbol: Zfp758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145484.2 | NP_663459.2 | 2 - 3 | 350 | 119 | |
| GenBank mRNA | BC022203.1 | - | 2 - 3 | 327 | 119 | |
| BC037455.1 | - | 2 - 3 | 351 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC037455.1, NM_145484.2 transcripts.
Set Membership: |
|

Zfp758
Genomic Map