Assay Details
| Gene Symbol: Rnf121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078509.1 | - | 9 | 1748 | 135 | |
| AK159032.1 | - | 9 | 1924 | 135 | ||
| AK159260.1 | - | 9 | 1814 | 135 | ||
| AK168821.1 | - | 9 | 1800 | 135 | ||
| AK169089.1 | - | 9 | 1800 | 135 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf121
Genomic Map