Assay Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010441.1 | - | 1 | 64 | 110 | |
| AK088367.1 | - | 1 | 57 | 110 | ||
| AK133839.1 | - | 1 | 65 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010441.1,AK088367.1,AK133839.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psmb5
Genomic Map