Assay Details
| Gene Symbol: Fam58b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197989.1 | NP_932106.1 | 1 | 748 | 113 | |
| GenBank mRNA | AK007413.1 | - | 1 | 696 | 113 | |
| BC027022.1 | - | 1 | 748 | 5367363 | 113 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam58b
Genomic Map