Assay Details
| Gene Symbol: Gpr182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007412.2 | NP_031438.2 | 2 | 1571 | 127 | |
| GenBank mRNA | AK031480.1 | - | 1506 | 127 | ||
| BY590401.1 | - | 1 | 356 | 127 | ||
| D17292.1 | - | 2 | 1465 | 127 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | D17292.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | AK031480.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | BB806742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | BY590401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | NM_007412.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | D17292.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | AK031480.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | BB806742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | BY590401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436068 | NM_007412.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205785 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 394 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14141 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112543 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102769 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154354 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147325 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | D17292.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | AK031480.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | BB806742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | BY590401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | NM_007412.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | D17292.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | AK031480.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | BB806742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | BY590401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436068 | NM_007412.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | D17292.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | AK031480.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | BB806742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | BY590401.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | NM_007412.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | D17292.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | AK031480.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | BB806742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | BY590401.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436068 | NM_007412.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537935 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147325 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154354 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102769 | - | - |
| Whole Mouse Genome G4122A | A_52_P620496 | AK031480.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | D17292.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | AK031480.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | BB806742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | BY590401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | NM_007412.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | D17292.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | AK031480.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | BB806742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | BY590401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436068 | NM_007412.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110194 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102769 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147325 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154354 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08709533 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08709539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08709521 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08698828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08833506 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08709524 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08709531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08833517 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08221684 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08221682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08833501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08619613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08619614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08271619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08709531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08781872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08334535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08365734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08781874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08709534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537927 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373329 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418554_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373325 | - | - |
| Mouse Genome 430 2.0 Array | 1418554_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418554_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11536_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11536_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa17540_at | - | - |
| Mouse Expression Set 430 | 1418554_PM_at | - | - |
| Mouse Expression Set 430 | 1418554_at | - | - |
| Murine Genome U74v2 Set | 162449_PM_f_at | - | - |
| Murine Genome U74v2 Set | 162449_f_at | - | - |
| Murine Genome U74v2 Set | 104271_at | - | - |
| Murine Genome U74v2 Set | 104271_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418554_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418554_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418554_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418554_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 803865 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2760014 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2900685 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002760014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900685 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2765378 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213615 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213615 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2765378 | - | - |
| mouse-6 v1.1 (Illumina) | scl011536.1_57-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0011536.1_68-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900685 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2760014 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl011536.1_57-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0011536.1_68-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760014 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900685 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2765378 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213615 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213615 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2765378 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl011536.1_57-S | - | - |

Gpr182
Genomic Map