Assay Details
| Gene Symbol: Olfr628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147097.2 | NP_667308.2 | 1 | 863 | 106 | |
| GenBank mRNA | BC109148.1 | - | 1 | 949 | 40046009 | 106 |
| BC109149.1 | - | 1 | 949 | 40046010 | 106 | |
| Gene Symbol: Olfr243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025386.1 | NP_001020557.1 | 1 | 863 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155859 | NM_001025386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184836 | NM_001025386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184836 | BC109149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184836 | NM_147097.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184836 | BC109148.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457643 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140853 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155859 | NM_001025386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184836 | NM_001025386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184836 | BC109149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184836 | NM_147097.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184836 | BC109148.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P232590 | BC109149.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P232590 | BC109148.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06307488 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140853 | - | - |
| Whole Mouse Genome G4122A | A_52_P232590 | BC109149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P232590 | BC109148.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06364157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06064124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06364177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06364181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06324234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25384570 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10555810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555814 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555809 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555809 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555811 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700012771.mm.1 | TC0700001505.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012771.mm.1 | NM_147097.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012771.mm.1 | NM_147097.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012770.mm.1 | TC0700001505.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012773.mm.1 | TC0700001505.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012772.mm.1 | TC0700001505.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 761226 | - | - |
| ABI Mouse Genome Survey Microarray | 875516 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6650750 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5690025 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5690025 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6650750 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006650750 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690025 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3163363 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2593666 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162805 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231661 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231661 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3162805 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3163363 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2593666 | - | - |
| mouse-6 v1.1 (Illumina) | scl0259101.2_195-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00259101.2_180-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_70794773-A | - | - |
| mouse-6 v1.1 (Illumina) | GI_70794773-I | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6650750 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690025 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00259101.2_180-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0259101.2_195-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650750 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690025 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3162805 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2593666 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3163363 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231661 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3163363 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2593666 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162805 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231661 | - | - |

Olfr628
Genomic Map