Assay Details
| Gene Symbol: Champ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181854.2 | NP_862902.1 | 2 | 2731 | 142 | |
| GenBank mRNA | AK132164.1 | - | 2 | 2694 | 142 | |
| AK133990.1 | - | 2 | 2657 | 142 | ||
| AK170835.1 | - | 2 | 2674 | 142 | ||
| BC028991.1 | - | 2 | 2643 | 5355869 | 142 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670891 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670894 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | AK170835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | BC028991.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | AK129444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | AK132164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | AK133990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | AK170835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | BC028991.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | AK129444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | AK132164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P382939 | AK133990.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937386 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318624 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 752047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61783 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28540 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119020 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117488 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | AK170835.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | BC028991.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | AK129444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | AK132164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | AK133990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | AK170835.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | BC028991.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | AK129444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | AK132164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P382939 | AK133990.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25670888 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117488 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06430933 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119020 | - | - |
| Whole Mouse Genome G4122A | A_52_P382939 | AK170835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382939 | BC028991.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382939 | AK129444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382939 | AK132164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382939 | AK133990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382939 | AK170835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382939 | BC028991.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382939 | AK129444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382939 | AK132164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382939 | AK133990.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115627 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119020 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117488 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06805041 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06761009 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06805037 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06761007 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07144882 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06805042 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06761008 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25670891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25671012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25670958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07347881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07292635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07347885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06820303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07379290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06783209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07394623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06782130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07077987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07144881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25670958 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10570430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570431 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436020_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454679_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443658_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570429 | - | - |
| Mouse Genome 430 2.0 Array | 1454679_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443658_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436020_at | - | - |
| Mouse Genome 430 2.0 Array | 1436020_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454679_at | - | - |
| Mouse Genome 430 2.0 Array | 1443658_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101994_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101994_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800001262.mm.1 | NM_181854.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001262.mm.1 | NM_181854.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001262.mm.1 | TC0800000133.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001261.mm.1 | NM_181854.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001261.mm.1 | NM_181854.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001261.mm.1 | TC0800000133.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001260.mm.1 | NM_181854.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001260.mm.1 | NM_181854.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001260.mm.1 | TC0800000133.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454679_PM_at | - | - |
| Mouse Expression Set 430 | 1443658_at | - | - |
| Mouse Expression Set 430 | 1436020_at | - | - |
| Mouse Expression Set 430 | 1443658_PM_at | - | - |
| Mouse Expression Set 430 | 1436020_PM_at | - | - |
| Mouse Expression Set 430 | 1454679_at | - | - |
| Murine Genome U74v2 Set | 135441_PM_at | - | - |
| Murine Genome U74v2 Set | 135441_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454679_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443658_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454679_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436020_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443658_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436020_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1450605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450605 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685309 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685309 | - | - |
| mouse-6 v1.1 (Illumina) | scl34046.2_46-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450605 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34046.2_46-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2685309 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2685309 | - | - |

Champ1
Genomic Map