Assay Details
| Gene Symbol: Fpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008042.2 | NP_032068.2 | 1 | 1177 | 111 | |
| GenBank mRNA | U78299.1 | - | 1225 | 111 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056205 | NM_008042.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056205 | U78299.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152941 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168630 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164694 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056205 | NM_008042.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056205 | U78299.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03288710 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08729343 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08729255 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08729261 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168630 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08729296 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03288740 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03288691 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164694 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08729235 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08729342 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164694 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168630 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08729296 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08729336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31075691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13585660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13585657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13404672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13506034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13394572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13394561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31075680 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442101 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421661_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450583_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442098 | - | - |
| Mouse Genome 430 2.0 Array | 1421661_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421661_at | - | - |
| Mouse Genome 430 2.0 Array | 1450583_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1450583_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14294_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14294_PM_at | - | - |
| Mouse Expression Set 430 | 1421661_at | - | - |
| Mouse Expression Set 430 | 1421661_PM_at | - | - |
| Mouse Expression Set 430 | 1450583_PM_s_at | - | - |
| Mouse Expression Set 430 | 1450583_s_at | - | - |
| Murine Genome U74v2 Set | 100741_PM_at | - | - |
| Murine Genome U74v2 Set | 95770_s_at | - | - |
| Murine Genome U74v2 Set | 95770_PM_s_at | - | - |
| Murine Genome U74v2 Set | 100741_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421661_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421661_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450583_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450583_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450583_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450583_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421661_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421661_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 545251 | - | - |
| ABI Mouse Genome Survey Microarray | 641351 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3360451 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3360451 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360451 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2754393 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221662 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2754393 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221662 | - | - |
| mouse-6 v1.1 (Illumina) | scl014294.2_322-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360451 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014294.2_322-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360451 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221662 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221662 | - | - |

Fpr3
Genomic Map