Assay Details
| Gene Symbol: Gpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008155.4 | NP_032181.2 | 16 | 1539 | 84 | |
| GenBank mRNA | AF108354.1 | - | 3 | 170 | 84 | |
| AK031081.1 | - | 16 | 1444 | 84 | ||
| AK076424.1 | - | 14 | 1195 | 84 | ||
| AK133827.1 | - | 16 | 1461 | 84 | ||
| AK134890.1 | - | 16 | 1473 | 84 | ||
| AK147124.1 | - | 16 | 1502 | 84 | ||
| AK150341.1 | - | 16 | 1470 | 84 | ||
| AK152508.1 | - | 11 | 843 | 84 | ||
| AK159838.1 | - | 16 | 1443 | 84 | ||
| AK169681.1 | - | 16 | 1468 | 84 | ||
| BC086640.1 | - | 16 | 1486 | 6832250 | 84 | |
| BC088995.1 | - | 16 | 1446 | 6403790 | 84 | |
| L09104.1 | - | 7 | 2678 | 84 | ||
| M14220.1 | - | 16 | 1468 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpi1
Genomic Map