Assay Details
| Gene Symbol: Igfals | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008340.3 | NP_032366.2 | 2 | 2065 | 106 | |
| GenBank mRNA | AK004926.1 | - | 2 | 2067 | 106 | |
| AK220290.1 | - | 2 | 2577 | 106 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | BC129875.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | BC129876.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | AK004926.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | AK220290.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | NM_008340.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | BC129875.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | BC129876.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | AK004926.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | AK220290.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277088 | NM_008340.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145726 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103151 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | BC129875.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | BC129876.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | AK004926.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | AK220290.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | NM_008340.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | BC129875.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | BC129876.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | AK004926.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | AK220290.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277088 | NM_008340.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | BC129875.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | BC129876.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | AK004926.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | AK220290.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | NM_008340.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | BC129875.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | BC129876.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | AK004926.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | AK220290.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P277088 | NM_008340.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100039 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103151 | - | - |
| Whole Mouse Genome G4122A | A_51_P277088 | BC129875.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277088 | BC129876.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277088 | AK004926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277088 | AK220290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277088 | NM_008340.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277088 | BC129875.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277088 | BC129876.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277088 | AK004926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277088 | AK220290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277088 | NM_008340.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112481 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13511700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13511701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13553395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100031 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442628 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422826_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442625 | - | - |
| Mouse Genome 430 2.0 Array | 1422826_at | - | - |
| Mouse Genome 430 2.0 Array | 1422826_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16005_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16005_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700002803.mm.1 | NM_008340.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002803.mm.1 | NM_008340.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002803.mm.1 | TC1700000355.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422826_PM_at | - | - |
| Mouse Expression Set 430 | 1422826_at | - | - |
| Murine Genome U74v2 Set | 97987_at | - | - |
| Murine Genome U74v2 Set | 162450_f_at | - | - |
| Murine Genome U74v2 Set | 97987_PM_at | - | - |
| Murine Genome U74v2 Set | 162450_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422826_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422826_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422826_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422826_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 611571 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6350195 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006350195 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222609 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222609 | - | - |
| mouse-6 v1.1 (Illumina) | scl016005.2_201-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6350195 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl016005.2_201-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1222609 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222609 | - | - |

Igfals
Genomic Map