Assay Details
| Gene Symbol: Ascl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008554.3 | NP_032580.2 | 2 | 1020 | 74 | |
| GenBank mRNA | AK010738.1 | - | 2 | 982 | 74 | |
| AK167324.1 | - | 2 | 1003 | 74 | ||
| BC019520.1 | - | 2 | 995 | 4163833 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ascl2
Genomic Map