Assay Details
| Gene Symbol: Polr2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009090.5 | NP_033116.2 | 9 | 1132 | 143 | |
| GenBank mRNA | AK146763.1 | - | 9 | 1062 | 143 | |
| AK151270.1 | - | 9 | 1060 | 143 | ||
| AK160053.1 | - | 9 | 1067 | 143 | ||
| BC002023.1 | - | 9 | 1050 | 3492868 | 143 | |
| BC099548.1 | - | 9 | 1077 | 30312083 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079858 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079861 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079853 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079860 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034230 | AK142621.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034230 | NM_053246.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034230 | AK160053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034230 | BC099548.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034230 | AK146763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239924 | AK160053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239924 | BC099548.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239924 | D83999.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239924 | AK146763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P239924 | AK151270.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096823 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940921 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 879914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617195 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167065 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130787 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239924 | AK160053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239924 | BC099548.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239924 | D83999.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239924 | AK146763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P239924 | AK151270.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034230 | AK142621.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034230 | NM_053246.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034230 | AK160053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034230 | BC099548.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034230 | AK146763.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239926 | AK160053.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239926 | BC002023.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239926 | BC099548.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239926 | D83999.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P239926 | AK146763.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079867 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167065 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130787 | - | - |
| Whole Mouse Genome G4122A | A_51_P239926 | AK160053.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239926 | BC002023.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239926 | BC099548.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239926 | D83999.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239926 | AK146763.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115133 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130787 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167065 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06959487 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07240806 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079858 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079861 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26079853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07243338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06718989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06852614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07025025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07238644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07238663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06707413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07238672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07165644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07025028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06707411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06852613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079873 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10384351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580783 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384350 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416341_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10574242 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10384349 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10580782 | - | - |
| Mouse Genome 430 2.0 Array | 1416341_at | - | - |
| Mouse Genome 430 2.0 Array | 1416341_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20021_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20021_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800008770.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008770.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008770.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008773.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008773.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008773.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008779.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008779.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008779.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008771.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008771.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008771.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008768.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008768.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008768.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008769.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008769.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008769.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008772.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008772.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008772.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008774.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008774.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008774.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008775.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008775.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008775.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008778.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008778.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008778.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008776.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008776.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008776.mm.1 | TC0800001124.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008777.mm.1 | NM_009090.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008777.mm.1 | NM_009090.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008777.mm.1 | TC0800001124.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416341_PM_at | - | - |
| Mouse Expression Set 430 | 1416341_at | - | - |
| Murine Genome U74v2 Set | 160146_PM_r_at | - | - |
| Murine Genome U74v2 Set | 160146_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416341_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416341_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416341_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416341_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 894345 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1690020 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001690020 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226263 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241168 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226263 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241168 | - | - |
| mouse-6 v1.1 (Illumina) | scl020021.9_309-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1690020 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl020021.9_309-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1690020 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226263 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226263 | - | - |

Polr2c
Genomic Map