Assay Details
| Gene Symbol: Ccr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009916.2 | NP_034046.2 | 2 | 1039 | 72 | |
| GenBank mRNA | AK042013.1 | - | 2 | 1041 | 72 | |
| BC117041.1 | - | 2 | 1045 | 40126360 | 72 | |
| BC119171.1 | - | 2 | 1044 | 8733920 | 72 | |
| X90862.1 | - | 2 | 1067 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
tolerance induction
chemotaxis
inflammatory response
immune response
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of positive chemotaxis
chemokine-mediated signaling pathway
signal transducer activity
G-protein coupled receptor activity
chemokine receptor activity
C-C chemokine receptor activity
neuron migration
tolerance induction
chemotaxis
inflammatory response
immune response
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of positive chemotaxis
chemokine-mediated signaling pathway
signal transducer activity
G-protein coupled receptor activity
chemokine receptor activity
C-C chemokine receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958215 | NM_009916.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958215 | AK042013.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189498 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839056 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144041 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172043 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958215 | NM_009916.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958215 | AK042013.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444476 | X90862.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444473 | X90862.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844938 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02035604 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172043 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144041 | - | - |
| Whole Mouse Genome G4122A | A_51_P444476 | X90862.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444473 | X90862.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128470 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172043 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144041 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07773837 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08022405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08110931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08161927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07851925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07864250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07942391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07584310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08160006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07929145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07762780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07851924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07929144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07630806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07773838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07846872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07851939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08022405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844986 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10597421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597423 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421655_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10597420 | - | - |
| Mouse Genome 430 2.0 Array | 1421655_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421655_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12773_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12773_at | - | - |
| Mouse Expression Set 430 | 1421655_a_at | - | - |
| Mouse Expression Set 430 | 1421655_PM_a_at | - | - |
| Murine Genome U74v2 Set | 94696_PM_at | - | - |
| Murine Genome U74v2 Set | 94696_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421655_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421655_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421655_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421655_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 618105 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480184 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480184 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480184 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2753620 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2753620 | - | - |
| mouse-6 v1.1 (Illumina) | scl35273.3_268-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480184 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35273.3_268-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480184 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2753620 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2753620 | - | - |

Ccr4
Genomic Map