Assay Details
| Gene Symbol: Ccr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009917.5 | NP_034047.2 | 2 | 2787 | 153 | |
| GenBank mRNA | AK154595.1 | - | 2 | 2788 | 153 | |
| AK155628.1 | - | 2 | 2762 | 153 | ||
| AK155632.1 | - | 2 | 2760 | 153 | ||
| AK171391.1 | - | 1246 | 153 | |||
| BG091923.1 | - | 1 | 153 | |||
| D83648.1 | - | 2 | 2750 | 153 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
chemotaxis
defense response
inflammatory response
immune response
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
positive regulation of gene expression
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
calcium-mediated signaling
positive regulation of cell-cell adhesion
negative regulation of cell migration
negative regulation of axon extension
positive regulation of fever generation
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
positive regulation of neuron differentiation
positive regulation of interleukin-1 beta secretion
positive regulation of inflammatory response
positive regulation of apoptotic process by virus
chemokine-mediated signaling pathway
response to cholesterol
neuron death
positive regulation of cell migration involved in sprouting angiogenesis
negative regulation of macrophage apoptotic process
negative regulation of neural precursor cell proliferation
glycoprotein binding
actin binding
signal transducer activity
G-protein coupled receptor activity
chemokine receptor activity
C-C chemokine receptor activity
protein kinase binding
C-C chemokine binding
chemokine (C-C motif) ligand 5 binding
calcium ion transport
chemotaxis
defense response
inflammatory response
immune response
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
positive regulation of gene expression
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
calcium-mediated signaling
positive regulation of cell-cell adhesion
negative regulation of cell migration
negative regulation of axon extension
positive regulation of fever generation
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
positive regulation of neuron differentiation
positive regulation of interleukin-1 beta secretion
positive regulation of inflammatory response
positive regulation of apoptotic process by virus
chemokine-mediated signaling pathway
response to cholesterol
neuron death
positive regulation of cell migration involved in sprouting angiogenesis
negative regulation of macrophage apoptotic process
negative regulation of neural precursor cell proliferation
glycoprotein binding
actin binding
signal transducer activity
G-protein coupled receptor activity
chemokine receptor activity
C-C chemokine receptor activity
protein kinase binding
C-C chemokine binding
chemokine (C-C motif) ligand 5 binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | D83648.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | AK170303.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | AK171391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | AK155632.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | AK154595.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | D83648.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | AK170303.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | AK171391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | AK155632.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578732 | AK154595.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207528 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161350 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | D83648.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | AK170303.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | AK171391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | AK155632.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | AK154595.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | D83648.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | AK170303.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | AK171391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | AK155632.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578732 | AK154595.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | D83648.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | AK170303.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | AK171391.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | AK155632.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | AK154595.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | D83648.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | AK170303.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | AK171391.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | AK155632.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P578732 | AK154595.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528873 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528891 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03810568 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528897 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528890 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528868 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528888 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528886 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528885 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03810564 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03810566 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528879 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03810575 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528894 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528884 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528883 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528871 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528896 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528877 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528880 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528900 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528875 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03810577 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528882 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528887 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03810581 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175558 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09528895 | - | - |
| Whole Mouse Genome G4122A | A_52_P578732 | D83648.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578732 | AK170303.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578732 | AK171391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578732 | AK155632.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578732 | AK154595.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578732 | D83648.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578732 | AK170303.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578732 | AK171391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578732 | AK155632.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578732 | AK154595.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103408 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03810566 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09528895 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03810581 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09528883 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03810575 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175558 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03810568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32899175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10598014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598017 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422260_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424727_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422259_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10598013 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10590635 | - | - |
| Mouse Genome 430 2.0 Array | 1422259_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422260_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1424727_at | - | - |
| Mouse Genome 430 2.0 Array | 1422260_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1424727_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422259_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12774_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12774_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900014089.mm.1 | NM_009917.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900014089.mm.1 | NM_009917.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900014089.mm.1 | TC0900001675.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422259_a_at | - | - |
| Mouse Expression Set 430 | 1424727_PM_at | - | - |
| Mouse Expression Set 430 | 1424727_at | - | - |
| Mouse Expression Set 430 | 1422260_x_at | - | - |
| Mouse Expression Set 430 | 1422259_PM_a_at | - | - |
| Mouse Expression Set 430 | 1422260_PM_x_at | - | - |
| Murine Genome U74v2 Set | 102719_f_at | - | - |
| Murine Genome U74v2 Set | 102718_at | - | - |
| Murine Genome U74v2 Set | 161968_f_at | - | - |
| Murine Genome U74v2 Set | 102718_PM_at | - | - |
| Murine Genome U74v2 Set | 102719_PM_f_at | - | - |
| Murine Genome U74v2 Set | 161968_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422260_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422259_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424727_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422260_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422259_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424727_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422259_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424727_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424727_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422260_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422260_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422259_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1980072 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5910722 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1980072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005910722 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001980072 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685392 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2725020 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2725020 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685392 | - | - |
| mouse-6 v1.1 (Illumina) | scl0012774.2_90-S | - | - |
| mouse-6 v1.1 (Illumina) | scl012774.5_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1980072 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5910722 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012774.5_9-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012774.2_90-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980072 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5910722 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2888191 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2685393 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2685392 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2685393 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2888191 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2685392 | - | - |

Ccr5
Genomic Map