Assay Details
| Gene Symbol: Srgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011157.2 | NP_035287.1 | 3 | 875 | 98 | |
| GenBank mRNA | AK151357.1 | - | 3 | 879 | 98 | |
| AK168124.1 | - | 3 | 877 | 98 | ||
| BC037076.1 | - | 3 | 865 | 5038568 | 98 | |
| X16133.1 | - | 3 | 875 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
protein processing
biomineral tissue development
mast cell secretory granule organization
T cell secretory granule organization
maintenance of protease location in mast cell secretory granule
maintenance of granzyme B location in T cell secretory granule
negative regulation of cytokine secretion
protein binding
collagen binding
apoptotic process
protein processing
biomineral tissue development
mast cell secretory granule organization
T cell secretory granule organization
maintenance of protease location in mast cell secretory granule
maintenance of granzyme B location in T cell secretory granule
negative regulation of cytokine secretion
protein binding
collagen binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094925 | M34603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094925 | X16133.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094925 | NM_011157.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094925 | AK087960.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094925 | AK151357.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405863 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492126 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150785 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139000 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150586 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159828 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094925 | M34603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094925 | X16133.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094925 | NM_011157.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094925 | AK087960.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094925 | AK151357.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27186429 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159828 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07080518 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07080509 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150586 | - | - |
| Whole Mouse Genome G4122A | A_51_P193019 | X16133.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193019 | NM_011157.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193019 | AK087960.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193019 | AK151357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193019 | AK157945.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125332 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101729 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159828 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150586 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08869391 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08869395 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08869387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08869393 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08869386 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08869396 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08869381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27186356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08692456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08214604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08430554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08396378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08869391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08541229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08618927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08630167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08261079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08535432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08605635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08209883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08267848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08801580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08869389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08290248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08910834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27186407 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10369619 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369616 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417426_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10369615 | - | - |
| Mouse Genome 430 2.0 Array | 1417426_at | - | - |
| Mouse Genome 430 2.0 Array | 1417426_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19073_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19073_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa10590_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000016120.mm.1 | NM_011157.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000016120.mm.1 | NM_011157.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000016120.mm.1 | TC1000002260.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417426_at | - | - |
| Mouse Expression Set 430 | 1417426_PM_at | - | - |
| Murine Genome U74v2 Set | 94085_at | - | - |
| Murine Genome U74v2 Set | 94085_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417426_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417426_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417426_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417426_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 927537 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4070239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4590632 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004590632 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004070239 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2751527 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2722732 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2722732 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2751527 | - | - |
| mouse-6 v1.1 (Illumina) | scl019073.1_109-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003786.1_57-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4070239 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4590632 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl019073.1_109-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003786.1_57-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4070239 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590632 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2751527 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2722732 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2722732 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2751527 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003786.1_57-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl019073.1_109-S | - | - |

Srgn
Genomic Map