Assay Details
| Gene Symbol: Ugt1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013701.3 | NP_038729.1 | 1 | 344 | 139 | |
| GenBank mRNA | AK002629.1 | - | 1 | 345 | 139 | |
| AY227195.1 | - | 1 | 354 | 139 | ||
| BC138676.1 | - | 1 | 343 | 8861697 | 139 | |
| BC145969.1 | - | 1 | 343 | 40131272 | 139 | |
| D87866.1 | - | 1 | 318 | 139 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ugt1a2
Genomic Map