Assay Details
| Gene Symbol: 4930407I02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019572.1 | - | 2 | 256 | 140 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ethanolamine kinase 1 | Etnk1 |
1110061E11Rik, 4930555L11Rik, AI195356, AI841245, C80956, D6Ertd3e, EKI1 |
-23.8kb | Anti-sense | Chr. 6 - 143166866 - 143208546 on NCBI Build GRCm38 |
| Downstream |
DNA segment, Chr 6, ERATO Doi 474, expressed |
D6Ertd474e |
1700001I24Rik, AU023129 |
13.1kb | Anti-sense | Chr. 6 - 143245888 - 143297879 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930407I02Rik
Genomic Map