Assay Details
| Gene Symbol: 1500012K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045814.1 | 3 | 526 | 143 | ||
| GenBank mRNA | AK035406.1 | - | 3 | 526 | 143 | |
More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC0700001084.mm
This assay targets the same location as RTA transcript cluster(s) TC0100001797.rn
This assay targets the same location as RTA transcript cluster(s) TC0100001797.rn
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P120047 | AK035406.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003103 | AK005235.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304647 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411129 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78469 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62638 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160791 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P120047 | AK035406.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003103 | AK005235.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160791 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06244591 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120047 | AK035406.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06612958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06340819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06612959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25239233 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0700009593.mm.1 | TC0700001084.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700009593.mm.1 | NR_045814.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009593.mm.1 | NR_045814.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009591.mm.1 | TC0700001084.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700009591.mm.1 | NR_045812.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009591.mm.1 | NR_045812.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009591.mm.1 | NR_045814.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009591.mm.1 | NR_045814.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009590.mm.1 | TC0700001084.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700009590.mm.1 | NR_045812.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009590.mm.1 | NR_045812.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009590.mm.1 | NR_045814.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009590.mm.1 | NR_045814.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102480131 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102480131 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2452330 | - | - |
| mouse-6 v1.1 (Illumina) | scl32526.3.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102480131 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102480131 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2452328 | - | - |

1500012K07Rik
Genomic Map