Assay Details
| Gene Symbol: 4833415N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI843905.1 | - | 1 | 106 | ||
| AK014704.1 | - | 2 | 911 | 106 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sorting nexin 29 | Snx29 |
4933437K13Rik, ENSMUSG00000079738, Gm11170, Gm1737, Gm930, Rundc2a |
-24.7kb | Sense | Chr. 16 - 11321256 - 11755931 on NCBI Build GRCm38 |
| Downstream | calcineurin-like phosphoesterase domain containing 1 | Cpped1 | C530044N13Rik | 22.2kb | Anti-sense | Chr. 16 - 11803684 - 11909475 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2320085 | AI843905.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2320085 | AK014704.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1019369 | AK014704.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30549078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30548979 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Murine Genome U74v2 Set | 135248_at | - | - |
| Murine Genome U74v2 Set | 135248_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 517412 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103520082 | - | - |

4833415N18Rik
Genomic Map